siRNA Details
| Gene Symbol: FAM117A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030802.3 | NP_110429.1 | 8 | 1148 | ||
| XM_017025182.1 | XP_016880671.1 | 8 | 1906 | |||
| GenBank mRNA | AF123073.1 | - | 8 | 1109 | ||
| AK302376.1 | - | 8 | 962 | |||
| BC037572.1 | - | 8 | 1160 | 5526752 | ||
| BC065199.1 | - | 8 | 1113 | 5214423 | ||

FAM117A
Genomic Map