siRNA Details
| Gene Symbol: TRIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030912.2 | NP_112174.2 | 3 | 875 | ||
| GenBank mRNA | AF220034.1 | - | 3 | 752 | ||
| AF281046.1 | - | 3 | 761 | |||
| BC021925.1 | - | 3 | 874 | 4857393 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM8| Gene Symbol: TRIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030912.2 | NP_112174.2 | 3 | 875 | ||
| GenBank mRNA | AF220034.1 | - | 3 | 752 | ||
| AF281046.1 | - | 3 | 761 | |||
| BC021925.1 | - | 3 | 874 | 4857393 | ||