siRNA Details
| Gene Symbol: TRIM56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030961.2 | NP_112223.1 | 3 | 349 | ||
| XM_011516589.2 | XP_011514891.1 | 2 | 201 | |||
| GenBank mRNA | BC005847.2 | - | 3 | 349 | 2907142 | |
| BC011882.2 | - | 3 | 266 | |||
| BC048194.1 | - | 3 | 283 | 5814702 | ||

TRIM56
Genomic Map