siRNA Details
| Gene Symbol: ST6GALNAC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320273.1 | NP_001307202.1 | 2 | 331 | ||
| NM_001320274.1 | NP_001307203.1 | 2 | 331 | |||
| NM_030965.2 | NP_112227.1 | 2 | 331 | |||
| GenBank mRNA | AJ507292.1 | - | 2 | 75 | ||
| AK056241.1 | - | 2 | 298 | |||
| AK095758.1 | - | 2 | 259 | |||
| AK295019.1 | - | 2 | 247 | |||
| AK300902.1 | - | 2 | 256 | |||
| BC001201.1 | - | 2 | 238 | 3356535 | ||
| DA166913.1 | - | 2 | 290 | |||
| DC358960.1 | - | 2 | 331 | |||

ST6GALNAC5
Genomic Map