siRNA Details
| Gene Symbol: CHST9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256316.1 | NP_001243245.1 | 5 | 572 | ||
| NM_031422.5 | NP_113610.2 | 6 | 610 | |||
| XM_005258362.3 | XP_005258419.1 | 2 | 158 | |||
| XM_017026033.1 | XP_016881522.1 | 5 | 509 | |||
| XM_017026034.1 | XP_016881523.1 | 7 | 733 | |||
| GenBank mRNA | AF239821.1 | - | 6 | 487 | ||
| AF332472.1 | - | 5 | 401 | |||
| AF332473.1 | - | 3 | 168 | |||
| AK093349.1 | - | 5 | 509 | |||
| AY358488.1 | - | 6 | 589 | |||
| BC025764.1 | - | 6 | 554 | 5205829 | ||
| BM919514.1 | - | 5 | 551 | |||
| KU178792.1 | - | 5 | 296 | |||
| KU178793.1 | - | 4 | 258 | |||

CHST9
Genomic Map