siRNA Details
| Gene Symbol: HIST1H3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003532.2 | NP_003523.1 | 1 | 174 | ||
| GenBank mRNA | CR542014.1 | - | 1 | 174 | ||
| Gene Symbol: HIST1H3I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK313905.1 | - | 2 | 534 | ||

HIST1H3E
Genomic Map