siRNA Details
| Gene Symbol: RIOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031480.2 | NP_113668.2 | 11, 12 | 1266 | ||
| NM_153005.1 | NP_694550.1 | 4, 5 | 576 | |||
| XM_005249446.3 | XP_005249503.1 | 11, 12 | 1311 | |||
| XM_011514933.2 | XP_011513235.2 | 11, 12 | 1228 | |||
| GenBank mRNA | AF258661.1 | - | 4, 5 | 576 | ||
| AK054568.1 | - | 11, 12 | 1257 | |||
| AK314467.1 | - | 11, 12 | 1295 | |||
| AL834277.1 | - | 10, 11 | 965 | |||
| BC006104.1 | - | 11, 12 | 1266 | 3537086 | ||

RIOK1
Genomic Map