siRNA Details
| Gene Symbol: GRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031485.3 | NP_113673.3 | 4 | 844 | ||
| GenBank mRNA | AF337808.1 | - | 4 | 623 | ||
| AK074676.1 | - | 4 | 812 | |||
| AK300225.1 | - | 3 | 462 | |||
| AK315625.1 | - | 4 | 636 | |||
| AL713729.1 | - | 4 | 619 | |||
| BC002440.2 | - | 4 | 619 | 3347366 | ||
| BC036281.1 | - | 4 | 595 | |||
| BC047351.1 | - | 4 | 595 | |||
| EF011614.1 | - | 4 | 617 | |||

GRWD1
Genomic Map