siRNA Details
| Gene Symbol: LONP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300948.1 | NP_001287877.1 | 12 | 2126 | ||
| NM_031490.3 | NP_113678.2 | 13 | 2258 | |||
| GenBank mRNA | AJ548761.1 | - | 13 | 2125 | ||
| AK027544.1 | - | 13 | 2091 | |||
| AK027666.1 | - | 9 | 1293 | |||
| AK074775.1 | - | 13 | 2123 | |||
| AK090452.1 | - | 1 | 674 | |||
| AK127125.1 | - | 13 | 1890 | |||
| AK130091.1 | - | 2 | 135 | |||
| AK172790.1 | - | 4 | 413 | |||
| AL834201.1 | - | 12 | 1624 | |||
| BC002916.2 | - | 1 | 94 | |||
| BC004234.2 | - | 1 | 94 | |||
| BC093910.1 | - | 13 | 2092 | 7939755 | ||
| BC093912.1 | - | 13 | 2092 | 7939757 | ||
| BC110434.1 | - | 9 | 1274 | |||
| BC143246.1 | - | 12 | 1960 | 9051747 | ||

LONP2
Genomic Map