siRNA Details
| Gene Symbol: USP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031907.1 | NP_114113.1 | 1 | 1222 | ||
| XM_017029892.1 | XP_016885381.1 | 2 | 1308 | |||
| GenBank mRNA | AF285593.1 | - | 1 | 1222 | ||
| AK292527.1 | - | 6 | 1640 | |||
| BC069073.1 | - | 1 | 1221 | 7216887 | ||
| BC101189.2 | - | 1 | 1220 | 40019498 | ||
| BC101190.2 | - | 1 | 1220 | 40019499 | ||
| BC101191.2 | - | 1 | 1220 | 40019500 | ||

USP26
Genomic Map