siRNA Details
| Gene Symbol: TMTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193451.1 | NP_001180380.1 | 4 | 739 | ||
| NM_175861.3 | NP_787057.2 | 4 | 830 | |||
| XM_005253499.4 | XP_005253556.1 | 4 | 805 | |||
| XM_017020003.1 | XP_016875492.1 | 4 | 806 | |||
| XM_017020004.1 | XP_016875493.1 | 4 | 805 | |||
| XM_017020005.1 | XP_016875494.1 | 4 | 804 | |||
| XM_017020006.1 | XP_016875495.1 | 4 | 809 | |||
| GenBank mRNA | AK127297.1 | - | 4 | 615 | ||
| BC042083.1 | - | 4 | 830 | 5749757 | ||
| GQ865601.1 | - | 4 | 739 | |||

TMTC1
Genomic Map