siRNA Details
| Gene Symbol: TSPAN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290212.1 | NP_001277141.1 | 3, 4 | 1016 | ||
| NM_031945.4 | NP_114151.3 | 2, 3 | 782 | |||
| GenBank mRNA | AB593081.1 | - | 3, 4 | 989 | ||
| AB593082.1 | - | 2, 3 | 777 | |||
| AF325213.1 | - | 2, 3 | 738 | |||
| BC032802.1 | - | 2, 3 | 753 | 5191718 | ||

TSPAN10
Genomic Map