siRNA Details
| Gene Symbol: OR3A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012373.2 | NP_036505.2 | 1 | 695 | ||
| XM_011524028.2 | XP_011522330.1 | 2 | 705 | |||
| GenBank mRNA | BC069415.1 | - | 1 | 695 | 7262255 | |
| BC108921.2 | - | 1 | 689 | 40023068 | ||

OR3A3
Genomic Map