siRNA Details
| Gene Symbol: HDAC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159286.1 | NP_001152758.1 | 13 | 1693 | ||
| NM_032019.5 | NP_114408.3 | 14 | 1753 | |||
| GenBank mRNA | AF393962.1 | - | 14 | 1380 | ||
| AF407272.1 | - | 14 | 1380 | |||
| AF407273.1 | - | 13 | 1320 | |||
| AF426160.1 | - | 14 | 1733 | |||
| AL512711.1 | - | 2 | 643 | |||
| BC032049.1 | - | 14 | 1566 | |||
| BC094734.1 | - | 12 | 1421 | |||
| BC125083.1 | - | 14 | 1427 | 40117772 | ||
| CR456465.1 | - | 14 | 1436 | |||

HDAC10
Genomic Map