siRNA Details
| Gene Symbol: HDAC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159286.1 | NP_001152758.1 | 17 | 1980 | ||
| NM_032019.5 | NP_114408.3 | 18 | 2040 | |||
| GenBank mRNA | AF393962.1 | - | 18 | 1667 | ||
| AF407272.1 | - | 18 | 1667 | |||
| AF407273.1 | - | 17 | 1607 | |||
| AF426160.1 | - | 18 | 2020 | |||
| AY450395.1 | - | 15 | 1432 | |||
| BC032049.1 | - | 18 | 1853 | |||
| BC094734.1 | - | 16 | 1708 | |||
| BC125082.1 | - | 12 | 1189 | |||
| BC125083.1 | - | 18 | 1714 | 40117772 | ||
| CR456465.1 | - | 18 | 1723 | |||

HDAC10
Genomic Map