siRNA Details
| Gene Symbol: PIP4K2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003559.4 | NP_003550.1 | 8 | 1474 | ||
| XM_011525326.2 | XP_011523628.1 | 9 | 1615 | |||
| XM_011525327.2 | XP_011523629.1 | 10 | 1134 | |||
| XM_011525330.2 | XP_011523632.1 | 7 | 810 | |||
| XM_017025196.1 | XP_016880685.1 | 9 | 1051 | |||
| XM_017025197.1 | XP_016880686.1 | 9 | 951 | |||
| XM_017025198.1 | XP_016880687.1 | 7 | 1437 | |||
| XM_017025199.1 | XP_016880688.1 | 8 | 889 | |||
| GenBank mRNA | BT019614.1 | - | 8 | 993 | ||
| U85245.1 | - | 8 | 1473 | |||

PIP4K2B
Genomic Map