siRNA Details
| Gene Symbol: KIRREL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032123.6 | NP_115499.5 | 3 | 362 | ||
| NM_199179.3 | NP_954648.3 | 2 | 212 | |||
| NM_199180.3 | NP_954649.3 | 3 | 504 | |||
| XM_011527362.1 | XP_011525664.1 | 4 | 564 | |||
| XM_011527363.1 | XP_011525665.1 | 4 | 545 | |||
| XM_011527365.1 | XP_011525667.1 | 3 | 414 | |||
| XM_011527366.1 | XP_011525668.1 | 2 | 675 | |||
| XM_017027348.1 | XP_016882837.1 | 3 | 1071 | |||
| GenBank mRNA | AB593116.1 | - | 3 | 468 | ||
| AK307865.1 | - | 3 | 825 | |||
| AL136654.1 | - | 3 | 487 | |||
| AY305301.1 | - | 3 | 306 | |||
| AY305302.1 | - | 2 | 156 | |||
| AY358742.1 | - | 3 | 490 | |||
| BC064925.1 | - | 5 | 487 | 6141879 | ||

KIRREL2
Genomic Map