siRNA Details
| Gene Symbol: UXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004182.3 | NP_004173.1 | 4 | 293 | ||
| NM_153477.2 | NP_705582.1 | 3 | 471 | |||
| NR_045560.1 | 4 | 293 | ||||
| GenBank mRNA | AF083241.1 | - | 4 | 259 | ||
| AF083242.1 | - | 3 | 409 | |||
| AF092737.1 | - | 4 | 274 | |||
| AK223320.1 | - | 4 | 247 | |||
| AK312072.1 | - | 4 | 259 | |||
| BC000720.1 | - | 3 | 228 | 3505415 | ||
| BC008890.1 | - | 3 | 228 | 3959862 | ||
| DA372561.1 | - | 4 | 293 | |||
| EL949569.1 | - | 4 | 271 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013840, BC082294, AK086822, AK163983, AK038495, and others.

UXT
Genomic Map