siRNA Details
| Gene Symbol: FAM175A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139076.2 | NP_620775.2 | 9 | 1149 | ||
| XM_005263280.4 | XP_005263337.1 | 10 | 1282 | |||
| XM_017008694.1 | XP_016864183.1 | 8 | 1055 | |||
| GenBank mRNA | AK021582.1 | - | 4 | 1357 | ||
| AK022704.1 | - | 9 | 1135 | |||
| AK023676.1 | - | 8 | 1049 | |||
| AY358576.1 | - | 9 | 1100 | |||
| BC039573.1 | - | 9 | 1096 | 6045433 | ||
| EF531340.1 | - | 9 | 1084 | |||

FAM175A
Genomic Map