siRNA Details
| Gene Symbol: RNASEH2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032193.3 | NP_115569.2 | 1, 2 | 357 | ||
| GenBank mRNA | AF312034.1 | - | 1, 2 | 196 | ||
| AF346606.1 | - | 1, 2 | 357 | |||
| AK024627.1 | - | 1, 2 | 217 | |||
| AK225131.1 | - | 1, 2 | 217 | |||
| BC023588.2 | - | 1, 2 | 237 | 4763204 | ||
| BM921525.1 | - | 1, 2 | 226 | |||

RNASEH2C
Genomic Map