siRNA Details
| Gene Symbol: MEX3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032246.4 | NP_115622.2 | 2 | 1134 | ||
| GenBank mRNA | AB095929.1 | - | 1 | 1946 | ||
| AK094145.1 | - | 1 | 367 | |||
| AK131424.1 | - | 2 | 1011 | |||
| AY950678.1 | - | 2 | 696 | |||
| BC036211.1 | - | 2 | 1132 | 5266259 | ||
| BC111545.1 | - | 2 | 1110 | 5742971 | ||

MEX3B
Genomic Map