siRNA Details
| Gene Symbol: SLC37A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287498.1 | NP_001274427.1 | 2 | 270 | ||
| NM_032295.3 | NP_115671.1 | 2 | 270 | |||
| NM_207113.2 | NP_996996.1 | 2 | 270 | |||
| XM_011516626.2 | XP_011514928.1 | 4 | 767 | |||
| XM_011516627.2 | XP_011514929.1 | 4 | 766 | |||
| XM_017012712.1 | XP_016868201.1 | 4 | 743 | |||
| XM_017012713.1 | XP_016868202.1 | 4 | 762 | |||
| XM_017012714.1 | XP_016868203.1 | 4 | 783 | |||
| XM_017012715.1 | XP_016868204.1 | 4 | 786 | |||
| XM_017012716.1 | XP_016868205.1 | 4 | 790 | |||
| XM_017012717.1 | XP_016868206.1 | 4 | 762 | |||
| XM_017012718.1 | XP_016868207.1 | 4 | 761 | |||
| GenBank mRNA | AK074823.1 | - | 2 | 231 | ||
| AK296543.1 | - | 2 | 224 | |||
| AK299276.1 | - | 2 | 214 | |||
| AK300265.1 | - | 2 | 228 | |||
| AL136583.1 | - | 2 | 177 | |||
| BC028380.1 | - | 2 | 220 | 4811814 | ||
| BC046567.1 | - | 2 | 222 | 5246993 | ||
| CD101739.1 | - | 2 | 278 | |||
| DB481623.1 | - | 2 | 270 | |||

SLC37A3
Genomic Map