siRNA Details
| Gene Symbol: CARD19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032310.4 | NP_115686.3 | 3, 4 | 524 | ||
| NR_134466.1 | 3, 4 | 535 | ||||
| NR_134467.1 | 4, 5 | 626 | ||||
| GenBank mRNA | BC004500.2 | - | 4, 5 | 393 | 3833318 | |
| BC091476.1 | - | 3, 4 | 378 | |||
| BX354102.2 | - | 4, 5 | 506 | |||
| DB550397.1 | - | 1, 2 | 20 | |||

CARD19
Genomic Map