siRNA Details
| Gene Symbol: VPS25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032353.3 | NP_115729.1 | 1, 2 | 91 | ||
| XM_017025224.1 | XP_016880713.1 | 1, 2 | 57 | |||
| GenBank mRNA | AB014763.1 | - | 1, 2 | 51 | ||
| AK312092.1 | - | 1, 2 | 78 | |||
| BC006282.2 | - | 1, 2 | 91 | 4025884 | ||

VPS25
Genomic Map