siRNA Details
| Gene Symbol: SARNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033082.3 | NP_149073.1 | 6 | 413 | ||
| NR_026722.1 | 5 | 366 | ||||
| NR_026723.1 | 6 | 413 | ||||
| GenBank mRNA | AF161434.1 | - | 6 | 380 | ||
| AF486281.1 | - | 6 | 358 | |||
| AK222719.1 | - | 6 | 391 | |||
| AK290508.1 | - | 6 | 381 | |||
| BC007099.1 | - | 6 | 379 | 4273903 | ||
| BC093051.1 | - | 5 | 332 | 30382893 | ||
| BI520421.1 | - | 6 | 413 | |||

SARNP
Genomic Map