siRNA Details
| Gene Symbol: ELOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032377.3 | NP_115753.1 | 4 | 287 | ||
| XM_017027356.1 | XP_016882845.1 | 5 | 442 | |||
| XM_017027357.1 | XP_016882846.1 | 4 | 301 | |||
| XM_017027358.1 | XP_016882847.1 | 4 | 441 | |||
| XM_017027359.1 | XP_016882848.1 | 4 | 357 | |||
| GenBank mRNA | AK001171.1 | - | 5 | 444 | ||
| BC007516.2 | - | 3 | 242 | 3028469 | ||
| BG718042.1 | - | 4 | 391 | |||

ELOF1
Genomic Map