siRNA Details
| Gene Symbol: C2orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032411.2 | NP_115787.1 | 4 | 541 | ||
| XM_006712799.3 | XP_006712862.1 | 5 | 560 | |||
| XM_011511993.2 | XP_011510295.1 | 4 | 478 | |||
| XM_017005106.1 | XP_016860595.1 | 4 | 460 | |||
| GenBank mRNA | AF325503.1 | - | 4 | 541 | ||
| AY358601.1 | - | 4 | 502 | |||
| BC021742.1 | - | 4 | 545 | 5260075 | ||

C2orf40
Genomic Map