siRNA Details
| Gene Symbol: CARD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324281.1 | NP_001311210.1 | 21 | 3137 | ||
| NM_032415.5 | NP_115791.3 | 20 | 3076 | |||
| XM_011515586.2 | XP_011513888.1 | 21 | 3105 | |||
| XM_011515587.2 | XP_011513889.1 | 20 | 3068 | |||
| GenBank mRNA | AF322641.1 | - | 20 | 2978 | ||
| AF352576.1 | - | 18 | 2651 | |||
| AK074049.1 | - | 20 | 2996 | |||
| BC111719.1 | - | 19 | 2762 | 40009168 | ||

CARD11
Genomic Map