siRNA Details
| Gene Symbol: ADGRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083909.2 | NP_001077378.1 | 7 | 1589 | ||
| NM_001291085.1 | NP_001278014.1 | 4 | 1090 | |||
| XM_005252695.1 | XP_005252752.1 | 16 | 3409 | |||
| XM_011540273.1 | XP_011538575.1 | 3 | 920 | |||
| GenBank mRNA | AB058731.1 | - | 4 | 1090 | ||
| AY181241.1 | - | 16 | 3409 | |||
| AY255564.1 | - | 3 | 973 | |||
| EF536972.1 | - | 6 | 1378 | |||
More Information
Set Membership: |
|

ADGRA1
Genomic Map