siRNA Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032453.1 | NP_115829.1 | 5 | 539 | ||
| XM_005259328.4 | XP_005259385.1 | 5 | 631 | |||
| XM_005259329.3 | XP_005259386.1 | 4 | 545 | |||
| XM_017027380.1 | XP_016882869.1 | 3 | 520 | |||
| XM_017027381.1 | XP_016882870.1 | 4 | 531 | |||
| GenBank mRNA | AB058732.1 | - | 2 | 272 | ||
| AK091585.1 | - | 4 | 434 | |||
| AK301135.1 | - | 5 | 535 | |||

ZNF527
Genomic Map