siRNA Details
| Gene Symbol: CNFN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032488.3 | NP_115877.2 | 3 | 209 | ||
| XM_005259332.2 | XP_005259389.1 | 4 | 314 | |||
| XM_011527396.1 | XP_011525698.1 | 4 | 309 | |||
| XM_011527397.1 | XP_011525699.1 | 4 | 255 | |||
| GenBank mRNA | AB049591.2 | - | 3 | 204 | ||
| AK312080.1 | - | 3 | 209 | |||
| BC101197.1 | - | 4 | 265 | 40021735 | ||
| BC101198.2 | - | 3 | 185 | 40021738 | ||

CNFN
Genomic Map