siRNA Details
| Gene Symbol: ST6GAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142351.1 | NP_001135823.1 | 4 | 1185 | ||
| NM_001142352.1 | NP_001135824.1 | 4 | 1255 | |||
| NM_001322362.1 | NP_001309291.1 | 4 | 1234 | |||
| NM_032528.2 | NP_115917.1 | 4 | 1255 | |||
| NR_136313.1 | 3 | 270 | ||||
| XM_006712802.1 | XP_006712865.1 | 4 | 1270 | |||
| XM_011511999.2 | XP_011510301.1 | 4 | 1367 | |||
| XM_011512000.2 | XP_011510302.1 | 4 | 1452 | |||
| XM_011512001.2 | XP_011510303.1 | 4 | 1452 | |||
| XM_017005110.1 | XP_016860599.1 | 4 | 1206 | |||
| GenBank mRNA | AB058780.1 | - | 4 | 1185 | ||
| AB059555.1 | - | 4 | 1241 | |||
| AJ512141.1 | - | 2 | 126 | |||
| AK307784.1 | - | 4 | 1255 | |||
| BC008680.1 | - | 4 | 1241 | 3856171 | ||

ST6GAL2
Genomic Map