siRNA Details
| Gene Symbol: TTBK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032538.1 | NP_115927.1 | 9 | 1076 | ||
| XM_011514947.2 | XP_011513249.1 | 8 | 866 | |||
| XM_017011364.1 | XP_016866853.1 | 8 | 866 | |||
| XM_017011365.1 | XP_016866854.1 | 8 | 866 | |||
| XM_017011366.1 | XP_016866855.1 | 8 | 866 | |||
| XM_017011367.1 | XP_016866856.1 | 8 | 866 | |||
| GenBank mRNA | AB058758.1 | - | 7 | 645 | ||
| AB218664.1 | - | 10 | 880 | |||
| AK131217.1 | - | 8 | 806 | |||

TTBK1
Genomic Map