siRNA Details
| Gene Symbol: PLA2G12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318124.1 | NP_001305053.1 | 2 | 394 | ||
| NM_032562.4 | NP_115951.2 | 2 | 394 | |||
| XM_005270244.4 | XP_005270301.1 | 2 | 619 | |||
| GenBank mRNA | AF339053.1 | - | 2 | 353 | ||
| AF349540.1 | - | 2 | 297 | |||
| AK313712.1 | - | 2 | 357 | |||
| AY358032.1 | - | 2 | 327 | |||
| BC093996.1 | - | 2 | 316 | 7939841 | ||
| BC111946.1 | - | 2 | 316 | 8327414 | ||
| BC143532.1 | - | 2 | 316 | 9052041 | ||
| BP274516.1 | - | 2 | 394 | |||

PLA2G12B
Genomic Map