siRNA Details
| Gene Symbol: DGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253891.1 | NP_001240820.1 | 2 | 438 | ||
| NM_032564.4 | NP_115953.2 | 3 | 567 | |||
| XM_011545304.2 | XP_011543606.1 | 3 | 271 | |||
| GenBank mRNA | AB048286.2 | - | 3 | 538 | ||
| AB073384.1 | - | 2 | 115 | |||
| AF384161.2 | - | 3 | 315 | |||
| AL834287.1 | - | 3 | 446 | |||
| AY358532.1 | - | 3 | 492 | |||
| AY780647.1 | - | 2 | 170 | |||
| BC015234.1 | - | 3 | 556 | 3903313 | ||

DGAT2
Genomic Map