siRNA Details
| Gene Symbol: DPY30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321209.1 | NP_001308138.1 | 5 | 386 | ||
| NM_032574.3 | NP_115963.1 | 5 | 526 | |||
| NR_135585.1 | 5 | 379 | ||||
| NR_135587.1 | 4 | 243 | ||||
| GenBank mRNA | AA442557.1 | - | 4 | 174 | ||
| AF226998.1 | - | 5 | 445 | |||
| AV725116.1 | - | 5 | 317 | |||
| BC015970.1 | - | 5 | 348 | 4081996 | ||
| BI752326.1 | - | 5 | 347 | |||
| BM505380.1 | - | 1 | 357 | |||
| BQ276899.1 | - | 386 | ||||

DPY30
Genomic Map