siRNA Details
| Gene Symbol: MGARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032623.3 | NP_116012.2 | 4 | 840 | ||
| GenBank mRNA | AF329088.1 | - | 4 | 727 | ||
| AF484960.1 | - | 4 | 840 | |||
| BC104173.1 | - | 4 | 726 | 40030666 | ||
| BC104174.1 | - | 4 | 726 | 40030667 | ||

MGARP
Genomic Map