siRNA Details
| Gene Symbol: PIGO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201484.1 | NP_001188413.1 | 12 | 2345 | ||
| NM_032634.3 | NP_116023.2 | 10 | 3498 | |||
| NM_152850.3 | NP_690577.2 | 11 | 1893 | |||
| XM_005251619.3 | XP_005251676.1 | 10 | 3123 | |||
| XM_017015222.1 | XP_016870711.1 | 11 | 3615 | |||
| XM_017015223.1 | XP_016870712.1 | 11 | 2274 | |||
| XM_017015224.1 | XP_016870713.1 | 11 | 2538 | |||
| GenBank mRNA | AK074064.1 | - | 5 | 4723 | ||
| AK090433.1 | - | 4 | 5927 | |||
| AL833956.1 | - | 10 | 3531 | |||
| AY358472.1 | - | 9 | 3104 | |||
| BC001030.2 | - | 8 | 1223 | |||
| BC013987.2 | - | 9 | 1309 | |||
| BC029271.1 | - | 9 | 2769 | |||
| BC036916.1 | - | 10 | 1858 | 5203414 | ||
| BC065282.1 | - | 12 | 2016 | 6172460 | ||

PIGO
Genomic Map