siRNA Details
| Gene Symbol: UNC119B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080533.2 | NP_001074002.1 | 1, 2 | 290 | ||
| GenBank mRNA | AK126367.1 | - | 1, 2 | 267 | ||
| DA324219.1 | - | 1, 2 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UNC119B| Gene Symbol: UNC119B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080533.2 | NP_001074002.1 | 1, 2 | 290 | ||
| GenBank mRNA | AK126367.1 | - | 1, 2 | 267 | ||
| DA324219.1 | - | 1, 2 | 291 | |||