siRNA Details
| Gene Symbol: ADGRG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308362.1 | NP_001295291.1 | 3 | 519 | ||
| NM_032787.2 | NP_116176.2 | 9 | 1343 | |||
| XM_011513245.2 | XP_011511547.1 | 8 | 1218 | |||
| GenBank mRNA | AK027360.1 | - | 9 | 1331 | ||
| AK131069.1 | - | 2 | 202 | |||
| AK172769.1 | - | 5 | 585 | |||
| AK172774.1 | - | 1 | 96 | |||
| AK295345.1 | - | 3 | 461 | |||
| AY181246.1 | - | 9 | 1075 | |||
| BC113451.1 | - | 9 | 1109 | 8322503 | ||
| BC130388.1 | - | 9 | 1109 | 40146419 | ||
More Information
Set Membership: |
|

ADGRG7
Genomic Map