siRNA Details
| Gene Symbol: AIFM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198696.1 | NP_001185625.1 | 8 | 1094 | ||
| NM_032797.5 | NP_116186.1 | 8 | 1091 | |||
| GenBank mRNA | AF337957.1 | - | 7 | 895 | ||
| AF506757.1 | - | 8 | 1091 | |||
| AK027403.1 | - | 8 | 970 | |||
| AK127353.1 | - | 8 | 2846 | |||
| BC006121.1 | - | 7 | 899 | 3506309 | ||
| BC023601.2 | - | 8 | 1078 | 4548458 | ||
| BX537621.1 | - | 5 | 1728 | |||

AIFM2
Genomic Map