siRNA Details
| Gene Symbol: MIGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032809.2 | NP_116198.2 | 11 | 1341 | ||
| XM_005252282.1 | XP_005252339.1 | 11 | 1360 | |||
| XM_011519117.1 | XP_011517419.1 | 11 | 1381 | |||
| XM_011519118.1 | XP_011517420.1 | 11 | 1290 | |||
| GenBank mRNA | AK027502.1 | - | 3 | 235 | ||
| AK074127.1 | - | 12 | 1457 | |||
| AK301241.1 | - | 8 | 880 | |||
| AK303036.1 | - | 11 | 1381 | |||
| BC009114.1 | - | 11 | 1341 | 4153683 | ||

MIGA2
Genomic Map