siRNA Details
| Gene Symbol: MIGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032809.2 | NP_116198.2 | 6 | 835 | ||
| XM_005252282.1 | XP_005252339.1 | 6 | 854 | |||
| XM_011519117.1 | XP_011517419.1 | 6 | 875 | |||
| XM_011519118.1 | XP_011517420.1 | 6 | 784 | |||
| GenBank mRNA | AK074127.1 | - | 6 | 830 | ||
| AK075421.1 | - | 6 | 828 | |||
| AK303036.1 | - | 6 | 875 | |||
| BC009114.1 | - | 6 | 835 | 4153683 | ||

MIGA2
Genomic Map