siRNA Details
| Gene Symbol: MAP4K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270425.1 | NP_001257354.1 | 13, 14 | 1321 | ||
| NM_003618.3 | NP_003609.2 | 13, 14 | 1321 | |||
| GenBank mRNA | AK291467.1 | - | 13, 14 | 1287 | ||
| AK314487.1 | - | 13, 14 | 1158 | |||
| BC071579.1 | - | 13, 14 | 1313 | 5296205 | ||

MAP4K3
Genomic Map