siRNA Details
| Gene Symbol: FAM104A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098832.1 | NP_001092302.1 | 3, 4 | 462 | ||
| NM_001289412.1 | NP_001276341.1 | 3, 4 | 235 | |||
| GenBank mRNA | BX537858.1 | - | 3, 4 | 366 | ||
| BX648968.1 | - | 4, 5 | 378 | |||
| HY026689.1 | - | 3, 4 | 235 | |||
| HY037907.1 | - | 3, 4 | 220 | |||
| U92989.1 | - | 3, 4 | 206 | |||

FAM104A
Genomic Map