siRNA Details
| Gene Symbol: SLC22A16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033125.3 | NP_149116.2 | 8 | 1704 | ||
| XM_005267184.3 | XP_005267241.1 | 8 | 1353 | |||
| XM_011536204.2 | XP_011534506.1 | 8 | 1718 | |||
| XM_011536205.2 | XP_011534507.1 | 8 | 1973 | |||
| XM_011536206.2 | XP_011534508.1 | 9 | 1406 | |||
| XM_011536207.2 | XP_011534509.1 | 8 | 1413 | |||
| XM_011536208.2 | XP_011534510.1 | 8 | 1326 | |||
| XM_011536209.2 | XP_011534511.1 | 9 | 1468 | |||
| XM_011536211.2 | XP_011534513.1 | 7 | 1208 | |||
| XM_011536212.2 | XP_011534514.1 | 6 | 1059 | |||
| GenBank mRNA | AB055798.1 | - | 10 | 1819 | ||
| AF268892.1 | - | 8 | 1637 | |||
| AI632467.1 | - | 1 | 290 | |||
| AK057306.1 | - | 7 | 1685 | |||
| AY145502.1 | - | 8 | 1637 | |||
| BC037247.1 | - | 8 | 1652 | |||
| BC047565.1 | - | 8 | 1704 | 4838011 | ||
| GQ472228.1 | - | 8 | 1704 | |||

SLC22A16
Genomic Map