siRNA Details
| Gene Symbol: TSLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033035.4 | NP_149024.1 | 4 | 620 | ||
| NM_138551.4 | NP_612561.2 | 2 | 402 | |||
| NR_045089.1 | 5 | 1825 | ||||
| XM_011543698.1 | XP_011542000.1 | 3 | 224 | |||
| GenBank mRNA | AF338732.1 | - | 4 | 454 | ||
| AL833421.1 | - | 2 | 382 | |||
| AY037115.1 | - | 4 | 620 | |||
| BC016720.1 | - | 2 | 335 | |||
| BC022894.1 | - | 2 | 374 | |||
| BC040592.1 | - | 5 | 1826 | 5269764 | ||
| BF245025.1 | - | 2 | 390 | |||
| BU571002.1 | - | 2 | 405 | |||

TSLP
Genomic Map