siRNA Details
| Gene Symbol: ASMTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173473.1 | NP_001166944.1 | 9 | 1206 | ||
| NM_001173474.1 | NP_001166945.1 | 8 | 1243 | |||
| NM_004192.3 | NP_004183.2 | 9 | 1291 | |||
| XM_005274434.3 | XP_005274491.1 | 10 | 1397 | |||
| XM_011546185.2 | XP_011544487.1 | 7 | 1039 | |||
| XM_017029919.1 | XP_016885408.1 | 8 | 1087 | |||
| GenBank mRNA | AK001090.1 | - | 9 | 1187 | ||
| AK090498.1 | - | 8 | 1195 | |||
| AK225770.1 | - | 9 | 1213 | |||
| AK297805.1 | - | 8 | 1124 | |||
| AK299231.1 | - | 9 | 1209 | |||
| AK301844.1 | - | 9 | 1200 | |||
| BC002508.2 | - | 9 | 1177 | 3051194 | ||
| BC010089.2 | - | 9 | 1175 | 2988425 | ||

ASMTL
Genomic Map