siRNA Details
| Gene Symbol: SSNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003731.2 | NP_003722.2 | 1, 2 | 171 | ||
| GenBank mRNA | BC000864.1 | - | 1, 2 | 126 | 3459939 | |
| BT006766.1 | - | 1, 2 | 46 | |||
| CR542238.1 | - | 1, 2 | 46 | |||
| DB466912.1 | - | 1, 2 | 171 | |||
| Z96932.1 | - | 1, 2 | 92 | |||

SSNA1
Genomic Map