siRNA Details
| Gene Symbol: KCNK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003740.3 | NP_003731.1 | 2, 3 | 651 | ||
| XM_005249456.1 | XP_005249513.1 | 2, 3 | 384 | |||
| GenBank mRNA | AF084830.1 | - | 2, 3 | 626 | ||
| AK314298.1 | - | 2, 3 | 384 | |||
| BC060793.1 | - | 2, 3 | 651 | 30348251 | ||
| BC069573.1 | - | 2, 3 | 358 | 7262361 | ||
| EU978936.1 | - | 2, 3 | 437 | |||

KCNK5
Genomic Map